3,5-Difluoro-4-isopropoxybenzaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
22496-1G
22496-25G
22496-500MG
22496-5G
76910-836EA
254.62
USD
76910-836
76910-838
76910-840
76910-842
3,5-Difluoro-4-isopropoxybenzaldehyde ≥97%
3,5-Difluoro-4-isopropoxybenzaldehyde
Formula:
C₁₀H₁₀F₂O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD21800655 CAS Number: 1509381-38-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...