(4-Bromo-2,6-difluorophenyl)trimethylsilane ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-2,6-difluoro-1-(trimethylsilyl)benzene

22464-1G 22464-25G 22464-500MG 22464-5G
76910-812EA 409.51 USD
76910-812 76910-814 76910-816 76910-818
(4-Bromo-2,6-difluorophenyl)trimethylsilane ≥97% research grade
(4-Bromo-2,6-difluorophenyl)trimethylsilane
Formula: C₉H₁₁BrF₂Si
Storage Temperature: Refrigerator
MDL Number: MFCD18642935
CAS Number: 184910-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR