3-Bromo-2-chloropyridin-4-ol ≥95%

Supplier: AOBChem USA

Warning

22041-1G 22041-25G 22041-500MG 22041-5G
76910-712EA 185.66 USD
76910-712 76910-714 76910-716 76910-718
3-Bromo-2-chloropyridin-4-ol ≥95%
3-Bromo-2-chloropyridin-4-ol
Formula: C₅H₃BrClNO
MW: 208.44 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18257581
CAS Number: 1211519-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR