3-Isopropoxy-5-trifluoromethylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

21957-1G 21957-25G 21957-5G
76910-664EA 96.55 USD
76910-664 76910-666 76910-668
3-Isopropoxy-5-trifluoromethylphenylboronic acid ≥97%
3-Isopropoxy-5-trifluoromethylphenylboronic acid
Formula: C₁₀H₁₂BF₃O₃
MW: 248.007 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16660265
CAS Number: 1256345-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR