3-Bromo-2-chloro-6-fluorobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (3-Bromo-2-chloro-6-fluorophenyl)methanol

21911-1G 21911-25G 21911-500MG 21911-5G
76910-642EA 260.99 USD
76910-642 76910-644 76910-646 76910-648
3-Bromo-2-chloro-6-fluorobenzyl alcohol ≥97%
3-Bromo-2-chloro-6-fluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₅BrClFO
MW: 239.469 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20923583
CAS Number: 1784628-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR