3-Bromo-6-isopropoxy-2-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

21881-1G 21881-25G 21881-5G
76910-628EA 84.88 USD
76910-628 76910-630 76910-632
3-Bromo-6-isopropoxy-2-methylpyridine ≥97%
3-Bromo-6-isopropoxy-2-methylpyridine
Formula: C₉H₁₂BrNO
MW: 230.102 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18434477
CAS Number: 1239611-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR