3-Bromo-2-iodophenol ≥95%

Supplier: AOBChem USA

Warning

21682-1G 21682-25G 21682-500MG 21682-5G
76910-574EA 124.13 USD
76910-574 76910-576 76910-578 76910-580
3-Bromo-2-iodophenol ≥95%
3-Bromo-2-iodophenol

Precaución: for Research Use Only

Formula: C₆H₄BrIO
Storage Temperature: Refrigerator
MDL Number: MFCD08166320
CAS Number: 855836-52-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR