3-Bromo-5-chloro-4-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

21640-1G 21640-25G 21640-500MG 21640-5G
76910-564EA 409.51 USD
76910-564 76910-566 76910-568 76910-570
3-Bromo-5-chloro-4-methylpyridine ≥97%
3-Bromo-5-chloro-4-methylpyridine
Formula: C₆H₅BrClN
Storage Temperature: Ambient
MDL Number: MFCD18415476
CAS Number: 1260010-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR