3-Chloro-6-fluoro-2-iodobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

21442-1G 21442-500MG 21442-5G
76910-506EA 952.69 USD
76910-506 76910-508 76910-510
3-Chloro-6-fluoro-2-iodobenzaldehyde ≥97%
3-Chloro-6-fluoro-2-iodobenzaldehyde
Formula: C₇H₃ClFIO
Storage Temperature: Refrigerator
CAS Number: 2288709-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR