3-Bromo-4-iodoisopropylbenzene ≥97%

Supplier: AOBChem USA

Warning

21339-1G 21339-25G 21339-5G
76910-484EA 62.6 USD
76910-484 76910-486 76910-488
3-Bromo-4-iodoisopropylbenzene ≥97%
3-Bromo-4-iodoisopropylbenzene
Formula: C₉H₁₀BrI
MW: 324.98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878133
CAS Number: 1000578-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR