4-Bromo-1-butoxy-2-chlorobenzene ≥97%

Supplier: AOBChem USA

Warning

21218-1G 21218-25G 21218-5G
76910-462EA 66.84 USD
76910-462 76910-464 76910-466
4-Bromo-1-butoxy-2-chlorobenzene ≥97%
4-Bromo-1-butoxy-2-chlorobenzene
Formula: C₁₀H₁₂BrClO
MW: 263.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09907662
CAS Number: 1266253-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR