4-(Butylcarbamoyl)-3-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

21205-1G 21205-25G 21205-5G
76910-450EA 63.66 USD
76910-450 76910-452 76910-454
4-(Butylcarbamoyl)-3-fluorophenylboronic acid ≥97%
4-(Butylcarbamoyl)-3-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₅BFNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD08689527
CAS Number: 874289-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR