4-Benzylcarbamoyl-3-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

21200-1G 21200-25G 21200-500MG 21200-5G
76910-434EA 130.5 USD
76910-434 76910-436 76910-438 76910-440
4-Benzylcarbamoyl-3-fluorophenylboronic acid ≥97%
4-Benzylcarbamoyl-3-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₃BFNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD08689501
CAS Number: 874289-19-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR