3-Fluoro-4-(piperidine-1-carbonyl)phenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

21194-1G 21194-25G 21194-5G
76910-406EA 65.78 USD
76910-406 76910-408 76910-410
3-Fluoro-4-(piperidine-1-carbonyl)phenylboronic acid ≥97% research grade
3-Fluoro-4-(piperidine-1-carbonyl)phenylboronic acid
Formula: C₁₂H₁₅BFNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD08436039
CAS Number: 874289-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR