3-Bromo-5-methoxymethylpyridine ≥97%

Supplier: AOBChem USA

Warning

21148-1G 21148-25G 21148-500MG 21148-5G
76910-372EA 320.4 USD
76910-372 76910-374 76910-376 76910-378
3-Bromo-5-methoxymethylpyridine ≥97%
3-Bromo-5-methoxymethylpyridine
Formula: C₇H₈BrNO
Storage Temperature: Refrigerator
MDL Number: MFCD17169857
CAS Number: 173999-17-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR