3-Bromo-2-(methylthio)pyridine ≥95% research grade
Supplier: AOBChem USA
|
Warning
|
21144-1G
21144-25G
21144-500MG
21144-5G
76910-358EA
135.8
USD
76910-358
76910-360
76910-362
76910-364
3-Bromo-2-(methylthio)pyridine ≥95% research grade
3-Bromo-2-(methylthio)pyridine
Formula:
C₆H₆BrNS Storage Temperature: Refrigerator |
MDL Number:
MFCD12964310 CAS Number: 51933-77-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...