3-Amino-6-bromo-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Bromo-1H-indazol-3-amine

21022-25G 21022-5G
76910-336EA 204.76 USD
76910-336 76910-338
3-Amino-6-bromo-1H-indazole ≥97%
3-Amino-6-bromo-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₆BrN₃
MW: 212.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05665872
CAS Number: 404827-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR