4-Bromo-2-(trifluoromethyl)benzoic acid ethyl ester ≥97%

Supplier: AOBChem USA

Warning

20984-1G 20984-25G 20984-5G
76910-326EA 109.28 USD
76910-326 76910-328 76910-330
4-Bromo-2-(trifluoromethyl)benzoic acid ethyl ester ≥97%
4-Bromo-2-(trifluoromethyl)benzoic acid ethyl ester
Formula: C₁₀H₈BrF₃O₂
MW: 297.069 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14698181
CAS Number: 1214378-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR