2-Methoxynicotinonitrile ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Cyano-2-methoxypyridine, 2-Methoxynicotinonitrile

20862-25G 20862-5G
76910-318EA 310.85 USD
76910-318 76910-320
2-Methoxynicotinonitrile ≥97%
2-Methoxynicotinonitrile 2-(4-Formylphenyl)isonicotinonitrile

Precaución: for Research Use Only

Formula: C₇H₆N₂O
MW: 134.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07363804
CAS Number: 7254-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR