3-Bromo-2-fluorophenol ≥97%
Supplier: AOBChem USA
|
Warning
|
20837-25G
20837-5G
76910-310EA
310.85
USD
76910-310
76910-312
3-Bromo-2-fluorophenol ≥97%
3-Bromo-2-fluorophenol
Precaución: for Research Use Only
Formula:
C₆H₄BrFO Storage Temperature: Ambient |
MDL Number:
MFCD09864673 CAS Number: 156682-53-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...