3-Bromo-2,6-difluorophenol ≥95%

Supplier: AOBChem USA

Warning

20683-1G 20683-25G 20683-5G
76910-276EA 102.91 USD
76910-276 76910-278 76910-280
3-Bromo-2,6-difluorophenol ≥95%
3-Bromo-2,6-difluorophenol

Precaución: for Research Use Only

Formula: C₆H₃BrF₂O
Storage Temperature: Refrigerator
MDL Number: MFCD12910487
CAS Number: 221220-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR