3-Bromo-(5-ethoxycarbonyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromo-5-(ethoxycarbonyl)phenylboronic acid

20655-25G 20655-5G
76910-266EA 403.15 USD
76910-266 76910-268
3-Bromo-(5-ethoxycarbonyl)phenylboronic acid ≥97%
3-Bromo-(5-ethoxycarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BBrO₄
Storage Temperature: Refrigerator
MDL Number: MFCD08689516
CAS Number: 913835-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR