3-Chloro-2-formylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

20574-1G 20574-25G 20574-5G
76910-244EA 117.76 USD
76910-244 76910-246 76910-248
3-Chloro-2-formylphenylboronic acid pinacol ester ≥97%
3-Chloro-2-formylphenylboronic acid pinacol ester
Formula: C₁₃H₁₆BClO₃
MW: 266.528 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18447610
CAS Number: 1352129-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR