3-Bromophenylboronic acid N-butyldiethanolamine ester ≥97%

Supplier: AOBChem USA

Warning

20515-1G 20515-25G 20515-5G
76910-220EA 78.51 USD
76910-220 76910-222 76910-224
3-Bromophenylboronic acid N-butyldiethanolamine ester ≥97%
3-Bromophenylboronic acid N-butyldiethanolamine ester
Formula: C₁₄H₂₁BBrNO₂
MW: 326.037 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09265136
CAS Number: 1257641-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR