3-Bromo-6-chloro-2-fluorobenzonitrile ≥97%

Supplier: AOBChem USA

Warning

20472-1G 20472-25G 20472-500MG 20472-5G
76910-204EA 236.59 USD
76910-204 76910-206 76910-208 76910-210
3-Bromo-6-chloro-2-fluorobenzonitrile ≥97%
3-Bromo-6-chloro-2-fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₂BrClFN
MW: 234.453 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11846056
CAS Number: 943830-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR