3-(Benzyloxy)-2-fluoro-5-(trifluoromethyl)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

20422-1G 20422-25G 20422-500MG 20422-5G
76910-164EA 186.72 USD
76910-164 76910-166 76910-168 76910-170
3-(Benzyloxy)-2-fluoro-5-(trifluoromethyl)benzoic acid ≥95%
3-(Benzyloxy)-2-fluoro-5-(trifluoromethyl)benzoic acid
Formula: C₁₅H₁₀F₄O₃
MW: 314.232 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631784
CAS Number: 2379321-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR