3-Formyl-5-methylbenzonitrile ≥97% research grade

Supplier: AOBChem USA

Warning

20412-1G 20412-25G 20412-5G
76910-150EA 114.58 USD
76910-150 76910-152 76910-154
3-Formyl-5-methylbenzonitrile ≥97% research grade
3-Formyl-5-methylbenzonitrile
Formula: C₉H₇NO
MW: 145.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18382518
CAS Number: 27633-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR