5-Bromo-m-toluonitrile ≥97%

Supplier: AOBChem USA

Warning

20410-25G 20410-5G
76910-138EA 310.85 USD
76910-138 76910-140
5-Bromo-m-toluonitrile ≥97%
5-Bromo-m-toluonitrile
Formula: C₈H₆BrN
Storage Temperature: Ambient
MDL Number: MFCD08061970
CAS Number: 124289-21-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR