(3-Hydroxy-5-methylphenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3-Hydroxy-5-methylphenylboronic acid

20337-1G 20337-25G 20337-500MG 20337-5G
76910-118EA 439.22 USD
76910-118 76910-120 76910-122 76910-124
(3-Hydroxy-5-methylphenyl)boronic acid ≥97% research grade
(3-Hydroxy-5-methylphenyl)boronic acid
Formula: C₇H₉BO₃
Storage Temperature: Refrigerator
MDL Number: MFCD16618932
CAS Number: 1256345-79-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR