3-Bromo-5-isopropoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

20316-1G 20316-25G 20316-5G
76910-072EA 64.72 USD
76910-072 76910-074 76910-076
3-Bromo-5-isopropoxybenzoic acid ≥97%
3-Bromo-5-isopropoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₃
Storage Temperature: Refrigerator
MDL Number: MFCD16618944
CAS Number: 1119779-04-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR