3,4,5-Trichlorophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3,4,5-Trichlorophenylboronic acid,pinacol ester, 4,4,5,5-Tetramethyl-2-(3,4,5-trichlorophenyl)-1,3,2-dioxaborolane

20303-25G 20303-5G
76910-054EA 328.88 USD
76910-054 76910-056
3,4,5-Trichlorophenylboronic acid pinacol ester ≥97%
3,4,5-Trichlorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₄BCl₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD11504973
CAS Number: 942069-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR