3-Fluoro-4-(methylthio)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

20282-25G 20282-5G
76910-040EA 466.8 USD
76910-040 76910-042
3-Fluoro-4-(methylthio)phenylboronic acid ≥97%
3-Fluoro-4-(methylthio)phenylboronic acid
Formula: C₇H₈BFO₂S
Storage Temperature: Ambient
MDL Number: MFCD12913934
CAS Number: 221030-80-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR