3-Carboxy-4-fluorophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid, 3-Carboxy-4-fluorobenzeneboronic acid pinacol ester

20257-25G 20257-5G
76910-034EA 283.27 USD
76910-034 76910-036
3-Carboxy-4-fluorophenylboronic acid pinacol ester ≥97%
3-Carboxy-4-fluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₆BFO₄
Storage Temperature: Ambient
MDL Number: MFCD12546519
CAS Number: 882679-10-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR