4-(Methoxymethyl)benzaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

20233-1G 20233-25G 20233-500MG 20233-5G
76910-018EA 245.07 USD
76910-018 76910-020 76910-022 76910-024
4-(Methoxymethyl)benzaldehyde ≥97% research grade
4-(Methoxymethyl)benzaldehyde
Formula: C₉H₁₀O₂
Storage Temperature: Refrigerator
MDL Number: MFCD11553601
CAS Number: 93943-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR