3-Hydroxy-4-methyl-benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Hydroxy-4-methylbenzaldehyde

20226-1G 20226-25G 20226-500MG 20226-5G
76910-002EA 132.62 USD
76910-002 76910-004 76910-006 76910-008
3-Hydroxy-4-methyl-benzaldehyde ≥97%
3-Hydroxy-4-methyl-benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₈O₂
MW: 136.148 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01632496
CAS Number: 57295-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR