3-Bromo-2-ethoxypyridine ≥97%

Supplier: AOBChem USA

Warning

20172-25G 20172-5G
76909-986EA 198.39 USD
76909-986 76909-988
3-Bromo-2-ethoxypyridine ≥97%
3-Bromo-2-ethoxypyridine

Precaución: for Research Use Only

Formula: C₇H₈BrNO
MW: 202.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00234309
CAS Number: 57883-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR