3-Bromo-2-propoxypyridine ≥97%

Supplier: AOBChem USA

Warning

20170-1G 20170-25G 20170-5G
76909-978EA 81.69 USD
76909-978 76909-980 76909-982
3-Bromo-2-propoxypyridine ≥97%
3-Bromo-2-propoxypyridine

Precaución: for Research Use Only

Formula: C₈H₁₀BrNO
Storage Temperature: Refrigerator
MDL Number: MFCD09743637
CAS Number: 760207-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR