3,4-Difluoro-(5-methoxycarbonyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3,4-Difluoro-5-(methoxycarbonyl)phenylboronic acid

20152-1G 20152-500MG 20152-5G
76909-954EA 857.21 USD
76909-954 76909-956 76909-958
3,4-Difluoro-(5-methoxycarbonyl)phenylboronic acid ≥97%
3,4-Difluoro-(5-methoxycarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₇BF₂O₄
Storage Temperature: Refrigerator
MDL Number: MFCD12025989
CAS Number: 1217500-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR