3-Fluoro-4-iodoanisole ≥97%

Supplier: AOBChem USA

Warning

20130-1G 20130-25G 20130-500MG 20130-5G
76909-938EA 203.7 USD
76909-938 76909-940 76909-942 76909-944
3-Fluoro-4-iodoanisole ≥97%
3-Fluoro-4-iodoanisole
Formula: C₇H₆FIO
MW: 252.025 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11226328
CAS Number: 458-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR