3,4-Difluoro-2-isopropoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

20090-1G 20090-25G 20090-500MG 20090-5G
76909-924EA 356.47 USD
76909-924 76909-926 76909-928 76909-930
3,4-Difluoro-2-isopropoxyphenylboronic acid ≥97%
3,4-Difluoro-2-isopropoxyphenylboronic acid
Formula: C₉H₁₁BF₂O₃
MW: 215.99 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17011989
CAS Number: 1451390-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR