3,5-Dibromo-2-fluorochlorobenzene ≥95%

Supplier: AOBChem USA

Warning

20053-1G 20053-25G 20053-5G
76909-910EA 79.57 USD
76909-910 76909-912 76909-914
3,5-Dibromo-2-fluorochlorobenzene ≥95%
3,5-Dibromo-2-fluorochlorobenzene
Formula: C₆H₂Br₂ClF
MW: 288.339 g/mol
Storage Temperature: Ambient
CAS Number: 1000573-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR