3,5-Dibromo-2-fluorobenzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

20049-1G 20049-25G 20049-500MG 20049-5G
76909-894EA 173.99 USD
76909-894 76909-896 76909-898 76909-900
3,5-Dibromo-2-fluorobenzoic acid ≥97% research grade
3,5-Dibromo-2-fluorobenzoic acid
Formula: C₇H₃Br₂FO₂
MW: 297.904 g/mol
Storage Temperature: Ambient
CAS Number: 183065-73-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR