3,5-Dichloro-4-ethoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

19912-1G 19912-500MG 19912-5G
76909-882EA 492.26 USD
76909-882 76909-884 76909-886
3,5-Dichloro-4-ethoxybenzoic acid ≥97%
3,5-Dichloro-4-ethoxybenzoic acid
Formula: C₉H₈Cl₂O₃
MW: 235.064 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03461923
CAS Number: 41490-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR