3-Ethoxy-2-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

19876-1G 19876-25G 19876-500MG 19876-5G
76909-846EA 257.8 USD
76909-846 76909-848 76909-850 76909-852
3-Ethoxy-2-fluorobenzaldehyde ≥97%
3-Ethoxy-2-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉FO₂
MW: 168.165 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24678362
CAS Number: 1204176-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR