3-Bromo-2-thiophenecarbonitrile ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromothiophene-2-carbonitrile

19821-25G 19821-5G
76909-836EA 431.79 USD
76909-836 76909-838
3-Bromo-2-thiophenecarbonitrile ≥95%
3-Bromo-2-thiophenecarbonitrile

Precaución: for Research Use Only

Formula: C₅H₂BrNS
MW: 188.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00068173
CAS Number: 18791-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR