3-Fluoro-2-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

19760-1G 19760-25G 19760-500MG 19760-5G
76909-808EA 155.96 USD
76909-808 76909-810 76909-812 76909-814
3-Fluoro-2-methoxyphenol ≥97%
3-Fluoro-2-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₇FO₂
Storage Temperature: Ambient
MDL Number: MFCD12756463
CAS Number: 96994-70-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR