(3-Methyl-4-(methylthio)phenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3-Methyl-4-(methylthio)phenylboronic acid

19736-1G 19736-25G 19736-500MG 19736-5G
76909-792EA 201.58 USD
76909-792 76909-794 76909-796 76909-798
(3-Methyl-4-(methylthio)phenyl)boronic acid ≥97% research grade
(3-Methyl-4-(methylthio)phenyl)boronic acid
Formula: C₈H₁₁BO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD28147715
CAS Number: 221031-07-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR