4-Bromo-2,5-dimethoxy-phenol ≥97%

Supplier: AOBChem USA

Warning

19577-1G 19577-25G 19577-500MG 19577-5G
76909-766EA 254.62 USD
76909-766 76909-768 76909-770 76909-772
4-Bromo-2,5-dimethoxy-phenol ≥97%
4-Bromo-2,5-dimethoxy-phenol

Precaución: for Research Use Only

Formula: C₈H₉BrO₃
Storage Temperature: Refrigerator
MDL Number: MFCD06409235
CAS Number: 557757-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR