4-Bromo-2,3-dimethylphenol ≥97%

Supplier: AOBChem USA

Warning

19506-25G 19506-5G
76909-746EA 418 USD
76909-746 76909-748
4-Bromo-2,3-dimethylphenol ≥97%
4-Bromo-2,3-dimethylphenol
Formula: C₈H₉BrO
MW: 201.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07778969
CAS Number: 22802-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR