3-Bromo-5-isopropylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

19480-1G 19480-25G 19480-5G
76909-722EA 64.72 USD
76909-722 76909-724 76909-726
3-Bromo-5-isopropylbenzoic acid ≥97%
3-Bromo-5-isopropylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₂
MW: 243.097 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20040502
CAS Number: 112930-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR